Skip to Content
Merck
All Photos(3)

Key Documents

454664

Sigma-Aldrich

2-Hydroxy-5-methylbenzaldehyde

98%

Synonym(s):

5-Methylsalicylaldehyde, 2,5-Cresotaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(CH3)CHO
CAS Number:
Molecular Weight:
136.15
Beilstein:
2040673
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

bp

217 °C (lit.)

mp

54-57 °C (lit.)

functional group

aldehyde

SMILES string

Cc1ccc(O)c(C=O)c1

InChI

1S/C8H8O2/c1-6-2-3-8(10)7(4-6)5-9/h2-5,10H,1H3

InChI key

ILEIUTCVWLYZOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Berelli Anupama et al.
Journal of fluorescence, 27(3), 953-965 (2017-01-18)
Ternary Cu(II) complexes [Cu(II)(L)(bpy)Cl] 1, [Cu(II)(L)(Phen)Cl] 2 [L = 2,3-dimethyl-1-phenyl-4(2 hydroxy-5-methyl benzylideneamino)-pyrazol-5-one, bpy = 2,2
Spectroscopy and stereochemistry of the optically active copper (II), cobalt (II) and nickel (II) complexes with Schiff bases N,N'-(1R,2R)-(-)-1, 2-cyclohexylenebis(3-methylbenzylideneiminato) and N,N'-(1R,2R)-(-)-1,2-cyclohexylenebis(5-methylbenzylideneiminato)
Szlyk E, et al.
Polyhedron, 21(27), 2711-2717 (2002)
Syntheses, structures and electrochemistry of copper (II) salicylaldehyde/tris (3-phenylpyrazolyl) borate complexes as models for the radical copper oxidases.
Halcrow MA, et al.
J. Chem. Soc., Dalton Trans., 11, 1753-1762 (1999)
Atsuko Masuya-Suzuki et al.
ChemistryOpen, 10(1), 46-55 (2021-01-22)
We prepared Ln(III) (Ln=Eu, Gd, and Yb) complexes with a tripodal Schiff base, tris[2-(5-methylsalicylideneimino)ethyl]amine (H3 L) and studied their photophysical properties. Upon ligand excitation, YbL showed Yb(III)-centered luminescence in the near-infrared region. While the overall quantum yield (0.60(1)%) of YbL
Bijoy Dey et al.
Chemistry, an Asian journal, 15(11), 1709-1721 (2020-04-18)
The reaction of Fe(NCS)3 prepared in situ in MeOH with 5-X-SalEen ligands (5-X-SalEen=condensation product of 5-substituted salicylaldehyde and N-ethylethylenediamine) provided three Fe(III) complexes, [Fe(5-X-SalEen)2 ]NCS; X=Me (1), X=Br (2), X=OMe (3). All the complexes reveal similar structural features but a very

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service