Skip to Content
Merck
All Photos(1)

Key Documents

D130605

Sigma-Aldrich

2,5-Dimethoxybenzaldehyde

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€43.50
100 G
€91.00

€43.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€43.50
100 G
€91.00

About This Item

Linear Formula:
(CH3O)2C6H3CHO
CAS Number:
Molecular Weight:
166.17
Beilstein:
509301
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€43.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

crystals

bp

146 °C/10 mmHg (lit.)

mp

46-48 °C (lit.)

SMILES string

[H]C(=O)c1cc(OC)ccc1OC

InChI

1S/C9H10O3/c1-11-8-3-4-9(12-2)7(5-8)6-10/h3-6H,1-2H3

InChI key

AFUKNJHPZAVHGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Exo,Endo Isomerism, MEP/DFT, XRD/HSA-Interactions of 2,5-Dimethoxybenzaldehyde: Explores the conformers of 2,5-Dimethoxybenzaldehyde using density functional theory and examines its thermal and optical behavior (Al-Zaqri et al., 2020).

  • Corrosion performance of Schiff base derived from 2, 5-dimethoxybenzyaldehyde: Discusses the synthesis of a Schiff base from 2,5-dimethoxybenzaldehyde and its application in corrosion resistance, supported by experimental and DFT studies (Eze et al., 2022).

  • Methoxamine synthesis in a biocatalytic 1-pot 2-step cascade approach: Describes the synthesis of Methoxamine from pyruvate and 2,5-dimethoxybenzaldehyde in a biocatalytic one-pot, two-step reaction, highlighting the efficiency and selectivity of the process (Erdmann et al., 2019).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service