Skip to Content
Merck
All Photos(1)

Key Documents

CLS2679638

Corning® polypropylene closure

orange

Synonym(s):

Corning closure

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€202.00

€202.00


Estimated to ship on17 April 2025



Select a Size

Change View
100 EA
€202.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

€202.00


Estimated to ship on17 April 2025


description

fits neck O.D. 37 mm

manufacturer/tradename

Corning 26796-38

color

orange

Looking for similar products? Visit Product Comparison Guide

General description

For culture tubes and flasks with long rimless necks. Position closure open for gas exhange or closed for humidified environment. Internal drip ring minimizes contamination.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service