Skip to Content
Merck
All Photos(1)

Key Documents

C73303

Sigma-Aldrich

2-Methylbenzyl chloride

99%

Synonym(s):

α-Chloro-o-xylene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€40.00

€40.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€40.00

About This Item

Linear Formula:
CH3C6H4CH2Cl
CAS Number:
Molecular Weight:
140.61
Beilstein:
386016
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€40.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.541 (lit.)

bp

197-199 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccccc1CCl

InChI

1S/C8H9Cl/c1-7-4-2-3-5-8(7)6-9/h2-5H,6H2,1H3

InChI key

VQRBXYBBGHOGFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service