Skip to Content
Merck
All Photos(3)

Key Documents

111961

Sigma-Aldrich

4-Chlorobenzyl chloride

95%

Synonym(s):

α,4-Dichlorotoluene, (4-Chlorophenyl)methyl chloride, 1-Chloro-4-(chloromethyl)benzene, NSC 34032

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€68.40
500 G
€96.10

€68.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€68.40
500 G
€96.10

About This Item

Linear Formula:
ClC6H4CH2Cl
CAS Number:
Molecular Weight:
161.03
Beilstein:
471558
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€68.40


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

solid

bp

216-222 °C (lit.)

mp

27-29 °C (lit.)

functional group

chloro

SMILES string

ClCc1ccc(Cl)cc1

InChI

1S/C7H6Cl2/c8-5-6-1-3-7(9)4-2-6/h1-4H,5H2

InChI key

JQZAEUFPPSRDOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chlorobenzyl chloride was the test compound for determining rat hepatocellular thiols by HPLC[1].
Reagent used in derivatization of, for example, a calix[4]arene[2] and a tryptamine.[3]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron, 50, 4087-4087 (1994)
Tonic inhibition of thermoregulation in the decerebrate monkey (Saimiri sciureus).
J C Liu
Experimental neurology, 64(3), 632-648 (1979-06-01)
Synthetic Communications, 24, 2247-2247 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service