Skip to Content
Merck
All Photos(2)

Documents

918571

Sigma-Aldrich

3D Printable Yttria-stabilized Zirconium(IV) Oxide Ink

greener alternative

Synonym(s):

3D printable ceramic ink, 3Y-ZrO2 ink, YSZ ink, Yttria stabilized zirconia ink, Yttrium stabilized zirconium oxide ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352119
NACRES:
NA.23

description

Printing methods compatible with: Direct-Ink-Writing 3D printing

Quality Level

form

paste

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

70 wt. % (Yttria stabilized zirconia)

color

white

particle size

120 nm

viscosity

2-25 Pa.s (25 °C at shear rate of 100 s-1)

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

This product can be used in material extrusion 3D printing technique.
This product is a thixotropic ink based on yttria stabilized zirconia (YSZ) nanoparticles. It is suitable for various direct ink write (aka robocasting) printing technologies . Our YSZ ink has also been validated for stereolithography (SLA) 3D printing. The ink offers a stable dispersion of YSZ, high viscosity, good printability, and long shelf life. Annealed patterns provide excellent chemical resistance and high surface area. The ink is suitable for applications such as membranes, catalysis, separators and reactors.

Caution

Handeling tips:
  • Store in an airtight sealed container in refrigerator.
  • Avoid exposure to heat.
  • Add the thermal initiator (product No.479896) to 0.3wt% of the total 3Y-ZrO2/PEGDA ink mass, and mix well before usage.

Preparation Note

Step-by-Step procedure for ink mixing and loading into syringe is in attached file.

Direct-Ink-Writing (DIW) Notes:
  • Syringe Nozzle sizes ranging from 250 μm − 800 μm can be used.
  • Initial pressure for DIW printing ranges from 12-18 psi, depending on nozzle diameter used.

Post Processing of DIW printed parts:
  • Curing @ 110°C for 12-16 hours.
  • Sintering using the following sequence:
Starting at RT, Ramp to 200°C for 6 hours.
Ramp to 300°C @ 1°C/min. Hold for 4 hours.
Ramp to 400°C @ 1°C/min. Hold for 2 hours.
Ramp to 800°C @ 2°C/min. Hold for 4 hours.
Ramp to 1090°C @ 2°C/min. Hold for 15 hours.
Ramp down to 50°C @ 2°C/min.

Storage and Stability

Tighly seal remaining ink, and store in 2-8° C fridge.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3D printing of ceramics: A review.
Chen Z, et al.
J. Eur. Ceram. Soc., 39(4), 661-687 (2019)
3D-printed nanoporous ceramics: Tunable feedstock for direct ink write and projection microstereolithography
Alyssa L. Troksa et al.
Materials & Design, 198 , 109337-109337 (2021)
Synthesis of Highly Porous Yttria-Stabilized Zirconia by Tape-Casting Methods.
Boaro M, et al.
Journal of the American Ceramic Society. American Ceramic Society, 395-400 (2004)

Articles

Additive manufacturing combined with nanomaterial-based inks enhances material properties, particularly in energy and environmental applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service