Skip to Content
Merck
All Photos(2)

Key Documents

921912

Sigma-Aldrich

3D Printable Ultra-High Temperature Boron Carbide Ink

greener alternative

Synonym(s):

3D Printable B4C Ink, Direct-Ink-Writable B4C Ink

Sign Into View Organizational & Contract Pricing

Select a Size

30 G
€60.90

€60.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
30 G
€60.90

About This Item

Empirical Formula (Hill Notation):
B4C
CAS Number:
Molecular Weight:
55.25
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€60.90


Check Cart for Availability

Request a Bulk Order

description

Printing methods compatible with: Direct-Ink-Writing 3D printing

Quality Level

form

paste

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

50 % (v/v) (Boron carbide)

color

gray

mean particle size

1.8 μm

viscosity

10-103 Pa.s (25 °C at 100 s-1)

greener alternative category

storage temp.

2-8°C

SMILES string

B21B3B4B2C431

InChI

1S/CB4/c2-1-3(2)5(1)4(1)2

InChI key

INAHAJYZKVIDIZ-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

This product can be used in material extrusion 3D printing technique.
This product is a thixotropic ink based on boron carbide (B4C) microparticles. It is suitable for various direct ink write (aka robocasting) printing technologies. The ink offers a stable dispersion of B4C, high viscosity, good printability, and long shelf life (when stored in an airtight sealed container)[1][2][3]. Sintered patterns provide excellent thermal stability and hardness. The ink is suitable for applications such as heat exchanges, high wear components, components in extreme temperature environments, etc.

Storage and Stability

Handeling tips:
Store in an airtight sealed container in refrigerator.
Avoid exposure to heat.


Step-by-Step procedure for ink mixing and loading into syringe is in attached file.


Storage:
Store in an airtight sealed container in refrigerator, do not freeze
Avoid exposure to heat
Mix well by hand or in a planetary mixer (preferred) before usage.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Additive manufacturing of graded B4C-Al cermets with complex shapes.
Chandrasekaran S, et al.
Materials & Design, 188, 108516-108516 (2020)
Complex shaped boron carbides from negative additive manufacturing.
Lu R, et al.
Materials & Design, 148, 8-16 (2018)
Boron Carbide: Structure, Properties, and Stability under Stress.
Domnich V, et al
Journal of the American Ceramic Society. American Ceramic Society, 94(11), 3605-3628 (2011)

Articles

Additive manufacturing combined with nanomaterial-based inks enhances material properties, particularly in energy and environmental applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service