Skip to Content
Merck
All Photos(2)

Key Documents

900695

Sigma-Aldrich

Graphene dispersion

greener alternative

forinkjet printing, photonically annealable

Synonym(s):

Conductive ink, Graphene ink, conductive ink

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€269.00

€269.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€269.00

About This Item

UNSPSC Code:
12141908
NACRES:
NA.23

€269.00


Check Cart for Availability

Request a Bulk Order

Product Name

Graphene ink, for inkjet printing, photonically annealable

Quality Level

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

2.2-3.4 wt. % (solid (graphene and ethyl cellulose) in cyclohexanone/terpineol)

resistivity

0.003-0.008 Ω-cm, thermally annealed 300 °C for 30 minutes, film thickness >100 nm

particle size

≤3 μm

surface tension

28-36 dyn/cm

viscosity

7-14 mPa.s(25 °C) (shear viscosity at 10 s-1)

bp

213-218 °C (Terpineol)

density

0.9-1.1 g/mL at 25 °C

greener alternative category

SMILES string

OC(C1=C2C(C3=C(C4=C(C5=C(C6=C(C7=C(C8=C(C9=C(C%10=C(C%11=C(C%12=C(C%13=C(C%14=C(C%15=CC=C%16O)C%16=C%17)C%17=C%18)C%18=C%19C(O)=O)C%19=C%20)C%20=C%21O)C%21=C%22)C%22=C%23)C%23=C%24O)C%24=C%25)C%25=C%26C(O)=O)C%26=C%27)C%27=C2)=C%28C%29=C(C%30=C%31C%28=C%3

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Graphene inks are highly conductive, low cost and super flexible. Click here for more information.

Application

Formulated with ethyl cellulose in cyclohexanone, terpineol, and diethylene glycol methyl ether for inkjet printing.
Curing condition: Photonic annealing, >4 J/cm2 incident (Xe flash lamp) (substrate dependent - most effective for low thermal conductivity/thermal mass substrates i.e., glass, polyimide, PEN, PET) or thermal annealing at 275-350 °C for 20-30 min.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

111.2 °F

Flash Point(C)

44 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Performance Solid-State Supercapacitors and Microsupercapacitors Derived from Printable Graphene Inks.
Li L, et al.
Advanced Energy Materials, 6(20) (2016)
Ethan B Secor et al.
Advanced materials (Deerfield Beach, Fla.), 27(42), 6683-6688 (2015-10-01)
Intense pulsed light (IPL) annealing of graphene inks is demonstrated for rapid post-processing of inkjet-printed patterns on various substrates. A conductivity of ≈25,000 S m(-1) is achieved following a single printing pass using a concentrated ink containing 20 mg mL(-1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service