Skip to Content
Merck
All Photos(1)

Documents

914495

Sigma-Aldrich

Polycaprolactone diacrylate

average Mn 5,000

Synonym(s):

PCL diacrylate, PCL-diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O2)nC10H14O5
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mn 5,000

color

off-white to beige

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Polycaprolactone is a biodegradable polymer commonly used in biomedical applications. It is a semicrystalline polymer that possesses relatively long degradation time. Polycaprolactone diacrylate carriers photoreactive end groups for crosslinking, which allows its application in 3D printing and controlled drug delivery research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maneesh Jaiswal et al.
Biomedical materials (Bristol, England), 5(6), 065014-065014 (2010-11-17)
A biodegradable semi-interpenetrating hydrogel network (semi-IPN) of polyacrylamide and gelatin was prepared using polycaprolactone diacrylate (mol. wt ∼ 640) as a crosslinker. The drug-polymer interaction and IPN formation were investigated by attenuated total reflectance-Fourier transform infrared (ATR-FTIR) and thermal gravimetric
Yinfeng He et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 105(6), 1645-1657 (2016-05-15)
A new type of photocrosslinkable polycaprolactone (PCL) based ink that is suitable for three-dimensional (3D) inkjet printing has been developed. Photocrosslinkable Polycaprolactone dimethylacrylate (PCLDMA) was synthesized and mixed with poly(ethylene glycol) diacrylate (PEGDA) to prepare an ink with a suitable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service