Skip to Content
Merck
All Photos(2)

Key Documents

802115

Sigma-Aldrich

Polycaprolactone dimethacrylate

average Mn 800

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€418.00

€418.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
€418.00

About This Item

Linear Formula:
(C6H10O2)nC12H18O5
UNSPSC Code:
12162002
NACRES:
NA.23

€418.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

mol wt

average Mn 800

refractive index

n20/D 1.471

density

1.094 g/mL at 25 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Polycaprolactone is a biodegradable polymer commonly used in biomedical applications. A light-activated Polycaprolactone dimethacrylate gel was used for the sustained delivery of an anti-VEGF antibody.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Puneet Tyagi et al.
Molecular pharmaceutics, 10(8), 2858-2867 (2013-06-06)
A light-activated polycaprolactone dimethacrylate (PCM) and hydroxyethyl methacrylate (HEMA) based gel network was developed to sustain the release of stable, active bevacizumab (an anti-VEGF antibody used to treat choroidal neovascularization) and used to assess sustained ex vivo delivery in rabbit

Articles

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service