929611
PCL-PEG-PCL diacrylate
PCL average Mn 800, PEG average Mn 4000
Synonym(s):
Acrylate-PCL-PEG-PCL-acrylate, Biodegradable triblock copolymer, PCL-b-PEG-b-PCL, PEG hydrogel
About This Item
Recommended Products
form
low-melting solid
Quality Level
mol wt
PCL Mn 800
PCL average Mn 800
PEG Mn 4000
PEG average Mn 4000
contains
≥50 ppm MEHQ
color
white to off-white
Ω-end
acrylate
α-end
acrylate
storage temp.
−20°C
SMILES string
O=C(OCCOC(CCCCCOC(C=C)=O)=O)CCCCCOC(C=C)=O
InChI
1S/C20H30O8/c1-3-17(21)25-13-9-5-7-11-19(23)27-15-16-28-20(24)12-8-6-10-14-26-18(22)4-2/h3-4H,1-2,5-16H2
InChI key
PCLZPCUCIICUMZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- PCL-PEG-PCL triblock copolymer
- Hydrolytically degradable PCL blocks
- Acrylate functional groups permit photopolymerization
Application
- Tissue engineering
- 3D printing
- Drug delivery
Features and Benefits
- PCL-PEG-PCL triblock copolymer
- Hydrolytically degradable PCL blocks
- Acrylate functional groups permit photopolymerization
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service