Skip to Content
Merck
All Photos(3)

Key Documents

912220

Sigma-Aldrich

NanoFabTx PLGA-micro

for synthesis of 1-5 μm particles

Synonym(s):

NanoFabTx reagent kit, PLGA microparticle kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€565.00

€565.00


Estimated to ship on24 March 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
€565.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.23

€565.00


Estimated to ship on24 March 2025


Request a Bulk Order

Quality Level

application(s)

advanced drug delivery

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The NanoFabTx PLGA-micro kit provides reagents (PLGA and stabilizer) and a protocol for microfluidic synthesis of microparticles between 1 to 5 μm. This reagent kit can be combined with the NanoFabTx Microfluidic - micro device kit (Cat.No. 911860) for microfluidic synthesis of PLGA micro particles.

Application

NanoFabTx PLGA-micro for synthesis of 1-5 μm particle reagent kit is designed for the synthesis of hydrophobic drug- encapsulated poly(lactic-co-glycolic acid) (PLGA) microparticles. The protocol gives users step-by-step instructions developed and tested by our formulation scientists. The protocol includes suggested conditions for incorporating drugs into the formulation and details how to tailor the parameters to yield 1 μm particles, 5 μm particles, or any size in between—all with low polydispersity. This kit affords scientists the synthetic control and reproducibility needed to encapsulate drugs, proteins, or other molecules with polymer microparticles of a chosen size and drug loading. With this kit, researchers can quickly identify the ideal microparticle size and drug loading for their research application. PLGA is a biocompatible and biodegradable polymer that has been widely used in drug delivery systems for controlled drug release of many different types of therapeutic molecules.

Features and Benefits

  • Flexible synthesis tool to create uniform and reproducible nanoparticles
  • Optimized to make microparticles 1-5 μm by microfluidics with low polydispersity
  • Enables microparticle formulation screening and optimization of microparticles 1-5 μm to select ideal particle size for enhanced drug loading and controlled release
  • Based on non-toxic, biodegradable polymers

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

NanoFabTx™ platform accelerates drug development with ready-to-use formulations and microfluidic devices for particle synthesis.

NanoFabTX kits enable precise drug delivery with lipid nanoparticles and liposomes for mRNA and nucleic acids.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service