Skip to Content
Merck
All Photos(1)

Key Documents

GF45989881

Polylactic Acid - Biopolymer

(PLA), granule, 3 mm nominal granule size, weight 100 g, natural

Synonym(s):

ME346310

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€276.00

€276.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
€276.00

About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

€276.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

granular

manufacturer/tradename

Goodfellow 459-898-81

weight

100 g

color

natural

nominal granule size

3 mm

General description

For updated SDS information please visit www.goodfellow.com

Application

  • Polylactic acid (PLA) biocomposite: Processing, additive manufacturing, and advanced applications: Discusses the enthusiasm towards manufacturing of biopolymers, particularly PLA, and its applications in various fields due to its favorable environmental and mechanical properties (RA Ilyas, SM Sapuan, MM Harussani, M Hakimi, 2021).
  • Synthesis and applications of biopolymer composites: Highlights the development and applications of biodegradable biopolymer composites and nanocomposites, with an emphasis on PLA due to its wide use and environmental benefits (AM Díez-Pascual, 2019).
  • Biopolymer blends based on poly (lactic acid): Focuses on the blending of PLA with other biopolymers to enhance its mechanical and thermal properties, vital for expanding its application scope in industrial and environmental sectors (R Al-Itry, K Lamnawar, A Maazouz, 2015).
  • Migration of oligomers from a food contact biopolymer based on polylactic acid (PLA): Examines the safety and chemical characteristics of PLA when used in food packaging, focusing on the migration of its components under various conditions and their compliance with food safety standards (S Ubeda, M Aznar, P Alfaro, C Nerín, 2019).

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Scaffaro et al.
Applied microbiology and biotechnology, 102(18), 7739-7756 (2018-07-17)
Poly(lactic acid)-based antimicrobial materials received considerable attention as promising systems to control microbial growth. The remarkable physicochemical properties of PLA such as renewability, biodegradability, and US Food and Drug Administration (FDA) approval for clinical use open up interesting perspectives for

Questions

1–6 of 6 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the ratio between PLLA and PDLA in this PLA product?

    1 answer
    1. Unfortunately, the ratio of Poly-L-Lactic acid to Poly-D-Lactic acid has not been determined.

      Helpful?

  4. At what temperature should it be dried?

    1 answer
    1. Typically GF45989881 is recommended to be dried at about 65 - 90 DegC.

      Helpful?

  5. What is the average Mw of this product

    1 answer
    1. The molecular weight of this material is 230,000 g/mol.

      Helpful?

  6. Does 1EA Packing of Polylactic Acid- Biopolymer (SKU -GF45989881-1EA) Contain 100 grams.

    1 answer
    1. Yes, as stated below the product name, this is a package of 100 grams.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service