908088
Platinum on carbon
Evonik Noblyst® P2065 3% Pt
Synonym(s):
Pt/C
About This Item
Recommended Products
form
powder
contains
~60% water
extent of labeling
3 wt. % loading (dry basis)
weight
Unit weight excludes weight of water
matrix
activated carbon support
SMILES string
[Pt]
InChI
1S/Pt
InChI key
BASFCYQUMIYNBI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- hydrogenation of aromatic nitro groups to hydroxylamines
- hydrogenation of aldehydes and ketones
Other Notes
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
nwg
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
An application guide to help determine which heterogeneous catalysts are best for different synthetic applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service