Skip to Content
Merck
All Photos(2)

Key Documents

330159

Sigma-Aldrich

Platinum on carbon

Evonik NOBLYST® P2056 5% Pt

Synonym(s):

Pt/C

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€145.00
50 G
€693.00

€145.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€145.00
50 G
€693.00

About This Item

Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€145.00


Check Cart for Availability

Request a Bulk Order

form

powder

contains

~50% water

reaction suitability

core: platinum
reagent type: catalyst

extent of labeling

5 wt. % loading (dry basis)

weight

Unit weight excludes weight of water

matrix

activated carbon, wet support

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Platinum on carbon (Pt/C) is a versatile catalyst used for the hydrogenation of alkenes, alkynes, nitro groups, ketones, and aromatics.[1] It is used for hydrogenolysis of cyclopropanes[2] and catalyzes the oxidation of alcohols.[3] It is a useful reagent for reductive couplings, avoids dehalogenation.[4]

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Legal Information

Noblyst is a registered trademark of Evonik Degussa GmbH

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of heterogeneous catalytic hydrogenation for organic synthesis (2001)
The hydrogenolysis of cyclopropane on a platinum stepped single crystal at atmospheric pressure.
Kahn D R, et al.
J. Catal., 34(2), 294-306 (1974)
Selective reductive coupling of nitro aliphatic compounds with aldehydes in hydrogen using gold catalyst.
Cisneros l, et al.
Chinese Journal of Catalysis, 37(10), 1756-1763 (2016)
Kaiser, H.-P.; Muchowski, J. M.
The Journal of Organic Chemistry, 49, 4203-4203 (1984)
Meurs, J. H. H.; Stopher, D. W.
Angewandte Chemie (International Edition in English), 28, 927-927 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service