Skip to Content
Merck
All Photos(1)

Key Documents

908126

Sigma-Aldrich

Palladium on alumina

Evonik Noblyst® P1159 5% Pd

Synonym(s):

Pd/Al2O3

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€199.00

€199.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€199.00

About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

€199.00


Check Cart for Availability

Request a Bulk Order

form

powder

contains

<3% water

extent of labeling

5 wt. % loading (dry basis)

weight

Unit weight excludes weight of water

matrix

alumina support

mp

1552 °C (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Evonik Noblyst® P1159 5% Pd is a palladium on alumina catalyst. It has been shown to be effective in the following reactions:
  • hydrogenation of aromatic nitro groups to hydroxylamines
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Other Notes

Unit weight is of net dry catalyst.

Legal Information

Manufactured by Evonik Degussa GmbH.
Noblyst is a registered trademark of Evonik Degussa GmbH

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

An application guide to help you select the most suitable heterogeneous catalysts for diverse synthetic applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service