Skip to Content
Merck
All Photos(2)

Key Documents

905070

Sigma-Aldrich

[(TEEDA)Ni(o-tolyl)Cl]

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€119.00

€119.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€119.00

About This Item

Empirical Formula (Hill Notation):
C17H31ClN2Ni
Molecular Weight:
357.59
UNSPSC Code:
12161600
NACRES:
NA.22

€119.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder or crystals

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst

mp

>300 °C

Application

Effective precatalyst for a variety of nickel-catalyzed transformations including cross-couplings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service