Skip to Content
Merck
All Photos(2)

Key Documents

911984

Sigma-Aldrich

(2,2′-Bipyridine)diiodonickel(II)

Synonym(s):

(bpy)NiI2

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€137.00

€137.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€137.00

About This Item

Empirical Formula (Hill Notation):
C10H8I2N2Ni
CAS Number:
Molecular Weight:
468.69
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€137.00


Check Cart for Availability

Request a Bulk Order

form

powder or crystals

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

>300 °C

SMILES string

[Ni]1([NH]2=C(C3=[NH]1C=CC=C3)C=CC=C2)(I)I

InChI key

HXZGEUKJJFSILG-UHFFFAOYSA-L

Application

(2,2′-Bipyridine)diiodonickel(II) is a catalyst for the cross-electrophile coupling of vinyl halides and alkyl halides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Muta. 2 - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kirk W Shimkin et al.
Journal of the American Chemical Society, 140(23), 7074-7078 (2018-05-26)
Nickel-catalyzed cross-electrophile couplings have recently emerged as highly effective and practical methods for the formation of C-C bonds. By merging this process with well-established π-π coupling chemistry, a new method for the synthesis of tetrasubstituted alkenes has been developed. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service