Skip to Content
Merck
All Photos(1)

Key Documents

912794

Sigma-Aldrich

Ni(COD)(DQ)

≥95%

Synonym(s):

Bis(1,5-cyclooctadiene)(duroquinone) nickel(0)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€95.30
1 G
€272.00

€95.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€95.30
1 G
€272.00

About This Item

Empirical Formula (Hill Notation):
C18H24NiO2
CAS Number:
Molecular Weight:
331.08
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

€95.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

parameter

temperature stable

mp

227 °C (decomposition)

SMILES string

[Ni](C)C.O=C2C(=C(C(=O)C(=C2C)C)C)C.C1CCC\C=C/CC1

InChI key

PRUJUYLPWIUASG-ZMKTYRQVSA-N

Application

Ni(COD)(DQ), as shown by the Engle lab, is an air- and thermally stable Ni(0) precatalyst. This 18-electron complex has a unique stability profile allowing for easy reaction setup outside an inert atmosphere. Transformations such as the Suzuki-Miyaura coupling, borylation of aryl halides, and alkene hydroarylation have been demonstrated.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Van T Tran et al.
Angewandte Chemie (International ed. in English) (2020-02-18)
We report that Ni(COD)(DQ) (COD=1,5-cyclooctadiene, DQ=duroquinone), an air-stable 18-electron complex originally described by Schrauzer in 1962, is a competent precatalyst for a variety of nickel-catalyzed synthetic methods from the literature. Due to its apparent stability, use of Ni(COD)(DQ) as a

Related Content

Engle lab pioneers efficient catalytic methods, leveraging substrate directivity for streamlined organic synthesis and selective product formation.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service