Skip to Content
Merck
All Photos(1)

Key Documents

901423

Sigma-Aldrich

ITIC-F

Synonym(s):

9-Bis(2-methylene-((3-(1,1-dicyanomethylene)-6,7-difluoro)-indanone))-5,5,11,11-tetrakis(4-hexylphenyl)-dithieno[2,3-d:2’,3’-d’]-s-indaceno[1,2-b:5,6-b’]dithiophene, IT-4F, ITIC-2F, ITIC-DF3

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€628.00

€628.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€628.00

About This Item

Empirical Formula (Hill Notation):
C94H78F4N4O2S4
CAS Number:
Molecular Weight:
1499.90
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

€628.00


Check Cart for Availability

Request a Bulk Order

description

Band gap: 1.52 eV

Quality Level

Assay

97%

form

solid

Orbital energy

HOMO -5.66 eV 
LUMO -4.14 eV 

SMILES string

Fc1cc2c(cc1F)C(=C(C#N)C#N)\C(=C\c3[s]c4c([s]c5c4C(c8c5cc9c(c8)c%10[s]c%11c([s]c(c%11)\C=C%14/C(=O)c%15c(cc(c(c%15)F)F)C/%14=C(C#N)C#N)c%10C9(c%13ccc(cc%13)CCCCCC)c%12ccc(cc%12)CCCCCC)(c7ccc(cc7)CCCCCC)c6ccc(cc6)CCCCCC)c3)\C2=O

InChI

1S/C94H78F4N4O2S4/c1-5-9-13-17-21-55-25-33-61(34-26-55)93(62-35-27-56(28-36-62)22-18-14-10-6-2)75-45-72-76(46-71(75)89-85(93)91-81(107-89)43-65(105-91)41-73-83(59(51-99)52-100)67-47-77(95)79(97)49-69(67)87(73)103)94(63-37-29-57(30-38-63)23-19-15-11-7-3,64

InChI key

JOZQXSUYCMNTCH-ODDCUFEPSA-N

Application

ITIC-F can be employed as the electron transport layer (ETL) in OFET devices. It can be blended with donor polymers to form the active layer in p-type OFETs. ITIC-F exhibits a broad absorption spectrum that extends into the visible and near-infrared regions. This characteristic allows for effective light absorption across a wide range of wavelengths and contributes to enhanced light-harvesting and power conversion efficiency in OPV devices. ITIC-F is commonly used as a non-fullerene acceptor in OPV devices. It serves as the electron acceptor material alongside a donor polymer in the photoactive layer.
ITIC-F is a derivative of ITIC possessing lower energy levels and a broader absorption spectrum. It is used as an n-type molecule for organic photovoltaics, allowing very high performances of over 13%.
ITIC-F is a non-fullerene acceptor molecule, which can be used in the fabrication of organic solar cells (OSCs) and organic photovoltaics (OPVs).[1][2][3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance limitations in thieno [3, 4-c] pyrrole-4, 6-dione-based polymer: ITIC solar cells
Yang F, et al.
Physical Chemistry Chemical Physics, 19(35), 23990-23998 (2017)
Efficient Non-Fullerene Organic Photovoltaic Modules Incorporating As-Cast and Thickness-Insensitive Photoactive Layers
Zhang T, et al.
Advanced Energy Materials, 8(25), 1801387-1801387 (2018)
Wenchao Zhao et al.
Journal of the American Chemical Society, 139(21), 7148-7151 (2017-05-18)
A new polymer donor (PBDB-T-SF) and a new small molecule acceptor (IT-4F) for fullerene-free organic solar cells (OSCs) were designed and synthesized. The influences of fluorination on the absorption spectra, molecular energy levels, and charge mobilities of the donor and
Highly efficient inverted ternary organic solar cells with polymer fullerene-free acceptor as a third component material
Zhang K, et al.
Journal of Power Sources, 413(35), 391-398 (2019)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service