Skip to Content
Merck
All Photos(1)

Key Documents

901097

Sigma-Aldrich

PDBT-T1

greener alternative

Synonym(s):

Poly[[5,10-bis(5-octyl-2-thienyl)dithieno[2,3-d:2′,3′-d′]benzo[1,2-b:4,5-b′]dithiophene-2,7-diyl]-2,5-thiophenediyl[5,7-bis(2-ethylhexyl)-4,8-dioxo-4H,8H-benzo[1,2-c:4,5-c′]dithiophene-1,3-diyl]-2,5-thiophenediyl]

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€444.00

€444.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€444.00

About This Item

Linear Formula:
(C72H78O2S10)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€444.00


Check Cart for Availability

Request a Bulk Order

description

Band gap: 1.93 eV

Quality Level

form

solid

mol wt

Mw 20,000-50,000 (GPC, PS standard)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>200 °C

solubility

soluble (hot o-dichlorobenzene)

Orbital energy

HOMO -5.36 eV 
LUMO -3.43 eV 

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Hole transport organic materials allow perfect energy level alignment with the absorber layer and therefore efficient charge collection, are prone to degradation in ambient conditions.Click here for more information.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuze Lin et al.
Advanced materials (Deerfield Beach, Fla.), 29(3) (2016-11-20)
Five polymer donors with distinct chemical structures and different electronic properties are surveyed in a planar and narrow-bandgap fused-ring electron acceptor (IDIC)-based organic solar cells, which exhibit power conversion efficiencies of up to 11%.
High-Performance Electron Acceptor with Thienyl Side Chains for Organic Photovoltaics.
Lin Y, et al.
Journal of the American Chemical Society, 138(14), 4955-4961 (2016)
Highly Efficient Parallel-?Like Ternary Organic Solar Cells.
Tao L, et al.
Chemistry of Materials, 29(7), 2914-2920 (2017)
Highly Efficient Parallel-Like Ternary Organic Solar Cells.
Tao L, et al.
Chemistry of Materials, 29(7), 2914-2920 (2017)
Yuze Lin et al.
Journal of the American Chemical Society, 138(14), 4955-4961 (2016-03-26)
We develop an efficient fused-ring electron acceptor (ITIC-Th) based on indacenodithieno[3,2-b]thiophene core and thienyl side-chains for organic solar cells (OSCs). Relative to its counterpart with phenyl side-chains (ITIC), ITIC-Th shows lower energy levels (ITIC-Th: HOMO = -5.66 eV, LUMO =

Articles

Novel biosensing devices, like bio-FET sensors, offer label-free analysis by detecting ionic and biomolecular charges, crucial for diagnostics and biological research.

Explore the eco-friendly potential of organic thin film transistors (OTFTs) for detecting chemical analytes, identifying viruses, and assisting in health diagnostics. This mini-review highlights challenges of achieving sustainability, safety, and biodegradability of each component of an OTFT sensor.

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service