Skip to Content
Merck
All Photos(2)

Key Documents

906387

Sigma-Aldrich

ITIC-Cl

Synonym(s):

3,9-bis(2-methylene-((3-(1,1-dicyanomethylene)-6,7-dichloro)-indanone))-5,5,11,11-tetrakis(4-hexylphenyl)-dithieno[2,3-d:2′,3′-d′]-s-indaceno[1,2-b:5,6-b′]dithiophene, ITIC-2Cl

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€319.00

€319.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€319.00

About This Item

Empirical Formula (Hill Notation):
C94H78Cl4N4O2S4
CAS Number:
Molecular Weight:
1565.72
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

€319.00


Check Cart for Availability

Request a Bulk Order

description

Band gap: 1.48 eV

Assay

98%

form

powder

Orbital energy

HOMO -5.68 eV 
LUMO -4.20 eV 

SMILES string

[s]1c2c([s]c(c2)\C=C%14/C(=O)c%15c(cc(c(c%15)Cl)Cl)C/%14=C(C#N)C#N)c3c1c4c(cc7c(c4)C(c%10c%11[s]c(cc%11[s]c%107)\C=C%12\C(=O)c%13c(cc(c(c%13)Cl)Cl)C\%12=C(C#N)C#N)(c9ccc(cc9)CCCCCC)c8ccc(cc8)CCCCCC)C3(c6ccc(cc6)CCCCCC)c5ccc(cc5)CCCCCC

InChI

1S/C94H78Cl4N4O2S4/c1-5-9-13-17-21-55-25-33-61(34-26-55)93(62-35-27-56(28-36-62)22-18-14-10-6-2)75-45-72-76(46-71(75)89-85(93)91-81(107-89)43-65(105-91)41-73-83(59(51-99)52-100)67-47-77(95)79(97)49-69(67)87(73)103)94(63-37-29-57(30-38-63)23-19-15-11-7-3,6

InChI key

CGRWDQBQNINDAH-WJLAHIJSSA-N

General description

ITCI-CI is a low band acceptor-donor-acceptor molecule with indacenodithieno[3,2‐b]thiophene (IT) as a central donor atom and 2‐(3‐oxo‐2,3‐dihydroinden‐1‐ylidene)malononitrile (IC) as an acceptor end group. It is also a non-fullerene electron acceptor with a push-pull structure that induces an intramolecular charge transfer and extends absorption.[1][2]

Application

ITCI-CI can be mainly used in the fabrication of polymeric solar cells with an efficiency that is less than 11% and excellent thermal stability.[1][2][3][4]
ITIC-Cl is a novel non-fullerene acceptor material designed to use in organic photovoltaic devices.ITIC-Cl is a derivative of the ITIC molecule, which is increasingly used to replace fullerene based derivatives for organic solar cells due to significantly improved performances and stability[5][6][7][3].Compared to ITIC and its other derivatives such as ITIC-F, ITIC-Cl has a narrower bandgap and lower energy levels, and thus will be more suitable to pair with various p-type polymers.ITIC-Cl is readily soluble in most organic solvents used to fabricate organic electronic devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Highly efficient inverted ternary organic solar cells with polymer fullerene-free acceptor as a third component material
Z. Kan, et al.
Journal of Power Sources, 413, 391-398 (2019)
A tetrachlorinated molecular non-fullerene acceptor for high performance near-IR absorbing organic solar cells
Laventure A and Welch GC
Journal of Material Chemistry C, 6(34), 9060-9064 (2018)
An electron acceptor challenging fullerenes for efficient polymer solar cells
Lin Y, et al.
Advanced Materials, 27(7), 1170-1174 (2015)
Liang Gao et al.
Advanced materials (Deerfield Beach, Fla.), 28(37), 8288-8295 (2016-07-09)
A nonfullerene polymer solar cell with a high efficiency of 9.26% is realized by using benzodithiophene-alt-fluorobenzotriazole copolymer J51 as a medium-bandgap polymer donor and the low-bandgap organic semiconductor ITIC with high extinction coefficients as the acceptor.
Wenchao Zhao et al.
Advanced materials (Deerfield Beach, Fla.), 28(23), 4734-4739 (2016-04-12)
A nonfullerene-based polymer solar cell (PSC) that significantly outperforms fullerene-based PSCs with respect to the power-conversion efficiency is demonstrated for the first time. An efficiency of >11%, which is among the top values in the PSC field, and excellent thermal

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service