Skip to Content
Merck
All Photos(2)

Key Documents

798983

Sigma-Aldrich

Graphene dispersion

greener alternative

for screen printing, with ethyl cellulose in terpineol, screen printable

Synonym(s):

Graphene ink, conductive ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Graphene ink, for screen printing, with ethyl cellulose in terpineol, screen printable

Quality Level

form

liquid

composition

Solid content, 13-20%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

0.003-0.008 Ω-cm, thermally annealed 300 °C for 30 minutes, film thickness >100 nm, 25 °C

particle size

≤3 μm

viscosity

5-50 Pa.s(25 °C) (shear viscosity at 10 s-1)

bp

213-218 °C (Terpineol)

density

0.9-1.1 g/mL at 25 °C

greener alternative category

SMILES string

OC(C1=C2C(C3=C(C4=C(C5=C(C6=C(C7=C(C8=C(C9=C(C%10=C(C%11=C(C%12=C(C%13=C(C%14=C(C%15=CC=C%16O)C%16=C%17)C%17=C%18)C%18=C%19C(O)=O)C%19=C%20)C%20=C%21O)C%21=C%22)C%22=C%23)C%23=C%24O)C%24=C%25)C%25=C%26C(O)=O)C%26=C%27)C%27=C2)=C%28C%29=C(C%30=C%31C%28=C%3

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". 3D printable inks help to design and print more complex designs with minimum waste and it is environmentally friendly. Click here for more information.

Application

Graphene dispersion formulated for screen printing.




Curing Condition: 250-350 °C, 20-30 min




The printed films are generally too thick for photonic annealing, so the substrate will need to be compatible with the thermal curing conditions (typically 250-300 C for 30 minutes works well for most applications). Suggested substrates include Si/SiO2 and polyimide.
This product can be used in material extrusion 3D printing technique.

Legal Information

Sold under Material Transfer Agreement with Mark Hersam group at Northwestern University.  

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and Versatile Photonic Annealing of Graphene Inks for Flexible Printed Electronics
Ethan B. Secor , Bok Y. Ahn , Theodore Z. Gao , Jennifer A. Lewis , and Mark C. Hersam
Advanced Materials, 27, 6683-6688 (2015)
High-Performance Solid-State Supercapacitors and Microsupercapacitors Derived from Printable Graphene Inks
Lei Li, Ethan B. Secor, Kan-Sheng Chen, Jian Zhu, Xiaolong Liu, Theodore Z. Gao
Advanced Energy Materials, 6, N/A-N/A (2016)

Articles

Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Graphene's unique properties spark interdisciplinary interest; its honeycomb structure offers electrical, optical, and mechanical marvels.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service