Skip to Content
Merck
All Photos(1)

Key Documents

901090

Sigma-Aldrich

Silver nanoparticles ink for screen printing

low curing temperature

Synonym(s):

Ag ink, Conductive ink, Smart’Ink S-CS21303 conductive ink

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€289.00

€289.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€289.00

About This Item

Linear Formula:
Ag
UNSPSC Code:
12141740
NACRES:
NA.23

€289.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

viscous liquid (paste)

viscosity

4,400-6,400 mPa.s(20 °C) (at 100 s-1)
6,400-10,000 mPa.s(20 °C) (at 40 s-1)

capacity

55±5 % loading (Nanoparticles loading)

storage temp.

2-8°C

General description

Product benefits:
  • Very high electrical conductivity.
  • Very low curing temperature for printing on flexible substrates.
  • Non CMR ink.
  • Unchanged conductivity on bending.
  • Compatible with various curing processes including photonic, IR, low vacuum oven and thermal.
  • Good adhesion on various flexible substrates (PET, Kapton®, …).
Smart′Ink S-CS21303 conductive ink developed for printed electronics market is particularly well suited for applications requiring high conductivity and high resolution at very low curing temperatures. This ink, based on silver, is perfectly adapted to design conductive tracks on flexible substrates and is suitable to produce antennas for IoT applications (HF, UHF).
This ink has been developed to address the applications which require very low curing temperatures.

Other Notes

Resistivity: ≤1 mΩ/sq, per 25 μm (screen printed on PET, curing at 200 °C/5 min)
≤3 mΩ/sq, per 25 μm (screen printed on PET, curing at 120 °C/5 min)
≤4 mΩ/sq, per 25 μm (screen printed on PET, curing at 80 °C/5 min)
≤5 mΩ/sq, per 25 μm (screen printed on PET, curing at 60 °C/5 min)

Preparation Note

Typical processing guideline:
  • Smart′Ink S-CS21303 conductive ink needs to be agitated before use.
  • Ready to use. No need to use thinner.

Legal Information

Product of Genes′Ink
Kapton is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service