Skip to Content
Merck
All Photos(1)

Key Documents

771422

Sigma-Aldrich

Vinyl acrylate

contains <600 ppm MEHQ as inhibitor

Synonym(s):

Divinyl ester, Ethenyl 2-propenoate, Ethylene acrylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€464.00

€464.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€464.00

About This Item

Linear Formula:
H2C=CHCO2CH=CH2
CAS Number:
Molecular Weight:
98.10
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€464.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

contains

<600 ppm MEHQ as inhibitor

refractive index

n20/D 1.432 (lit.)
n20/D 1.432

bp

90-92 °C (lit.)

density

0.942 g/mL at 25 °C (lit.)
0.947 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=COC(=O)C=C

InChI

1S/C5H6O2/c1-3-5(6)7-4-2/h3-4H,1-2H2

InChI key

BLCTWBJQROOONQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This acrylate monomer has a vinyl group for functionalization. It can also be used as a crosslinkable monomer.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy M O'Shea et al.
Advanced healthcare materials, 4(12), 1802-1812 (2015-06-20)
Hydrogels with covalently incorporated trehalose are synthesized using thiol-ene Michael addition. Trehalose hydrogels afford prolonged stabilization and -controlled release of model enzymes in vitro and in vivo as well as preservation of protein stability under heat and -lyophilization stressors. Strong

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service