Skip to Content
Merck
All Photos(3)

Key Documents

235768

Sigma-Aldrich

Vinyltrimethoxysilane

98%

Synonym(s):

(Trimethoxysilyl)ethylene, Ethenyltrimethoxysilane, Trimethoxy(vinyl)silane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€79.80
500 ML
€204.00

€79.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€79.80
500 ML
€204.00

About This Item

Linear Formula:
H2C=CHSi(OCH3)3
CAS Number:
Molecular Weight:
148.23
Beilstein:
1099136
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€79.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.392 (lit.)

bp

123 °C (lit.)

density

0.968 g/mL at 25 °C (lit.)

SMILES string

CO[Si](OC)(OC)C=C

InChI

1S/C5H12O3Si/c1-5-9(6-2,7-3)8-4/h5H,1H2,2-4H3

InChI key

NKSJNEHGWDZZQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vinyltrimethoxysilane (VTMS) is a silane coupling agent with a silicon and hydroxyl groups that can be used to enhance the wettability and improve the superhydrophobic characteristics of different composites.[1][2]

Application

VTMS may be used to provide superhydrophobicity to different materials like TiO2, talc, kaolin, magnesium oxide nanoparticles, ammonium phosphate and PEDOT. It modifies the surface by capping the material and creates a protective layer that is water resistant and can be used in major coating industries.[1][2][3][4][5][6]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadir Hussain et al.
Nanomaterials (Basel, Switzerland), 11(2) (2021-03-07)
Plastic bottles are generally recycled by remolding them into numerous products. In this study, waste from plastic bottles was used to fabricate recycled polyethylene terephthalate (r-PET) nanofibers via the electrospinning technique, and high-performance conductive polyethylene terephthalate nanofibers (r-PET nanofibers) were
Mariana E Ghica et al.
Polymers, 12(6) (2020-06-07)
The present work describes for the first time the preparation of silica-based aerogel composites containing tetraethoxysilane (TEOS) and vinyltrimethoxysilane (VTMS) reinforced with Kevlar® pulp. The developed system was extensively investigated, regarding its physical, morphological, thermal and mechanical features. The obtained
Synthesis and characterization of PEDOT: P (SS-co-VTMS) with hydrophobic properties and excellent thermal stability.
Cho W, et al.
Polymer, 8(5), 189-189 (2016)
UV-induced switching behavior of novel fluoroalkyl end-capped vinyltrimethoxysilane oligomer/titanium oxide nanocomposite between superhydrophobicity and superhydrophilicity with good oleophobicity.
Sawada E, et al.
Composites Part B: Engineering, 41(6), 498-502 (2010)
Preparation and surface property of fluoroalkyl end-capped vinyltrimethoxysilane oligomer/talc composite-encapsulated organic compounds: application for the separation of oil and water.
Oikawa Y, et al.
ACS Applied Materials & Interfaces, 7(25), 13782-13793 (2015)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service