Skip to Content
Merck
All Photos(1)

Key Documents

139548

Sigma-Aldrich

Di(ethylene glycol) divinyl ether

99%

Synonym(s):

3,6,9-Trioxaundeca-1,10-diene, Bis[2-(vinyloxy)ethyl] ether, DEGDVE

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€230.00

€230.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
€230.00

About This Item

Linear Formula:
(H2C=CHOCH2CH2)2O
CAS Number:
Molecular Weight:
158.19
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€230.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

refractive index

n20/D 1.446 (lit.)

bp

198-199 °C (lit.)

density

0.968 g/mL at 25 °C (lit.)

SMILES string

C=COCCOCCOC=C

InChI

1S/C8H14O3/c1-3-9-5-7-11-8-6-10-4-2/h3-4H,1-2,5-8H2

InChI key

SAMJGBVVQUEMGC-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service