Skip to Content
Merck
All Photos(2)

Key Documents

757861

Sigma-Aldrich

Poly(ethylene glycol) 2-aminoethyl ether acetic acid

average Mn 1,100, cross-linking reagent amine reactive, cross-linking reagent carboxyl reactive

Synonym(s):

Polyethylene glycol, Amine-PEG-carboxylic acid, Amino-PEG acetic acid, AminoPEG carboxylic acid, Aminopolyoxyethylene acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€320.00

€320.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€320.00

About This Item

Linear Formula:
H2NC2H4O(C2H4O)nCH2CO2H
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€320.00


Check Cart for Availability

Request a Bulk Order

Product Name

Poly(ethylene glycol) 2-aminoethyl ether acetic acid, average Mn 1,100

form

solid

Quality Level

mol wt

PEG average Mn 1,000 (n~22)
average Mn 1,100

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

reagent type: cross-linking reagent
reactivity: carboxyl reactive

transition temp

Tm 48-55 °C

Ω-end

amine

α-end

carboxylic acid

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service