Skip to Content
Merck
All Photos(1)

Key Documents

JKA12008

Sigma-Aldrich

HO-PEG-NH2

hydroxyl PEG8 amine, monodisperse

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€381.00

€381.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€381.00

About This Item

Linear Formula:
HO(CH2CH2O)7CH2CH2NH2
UNSPSC Code:
51171641
NACRES:
NA.23

€381.00


Check Cart for Availability

Request a Bulk Order

Product Name

HO-PEG8-NH2,

form

solid

storage temp.

−20°C

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service