Skip to Content
Merck
All Photos(1)

Key Documents

749443

Sigma-Aldrich

L-Ornithine-3,3,4,4,5,5-d6 hydrochloride

≥98 atom % D, ≥98% (CP)

Synonym(s):

(S)-(+)-2,5-Diaminopentanoic acid-3,3,4,4,5,5-d6 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€358.00

€358.00


Check Cart for Availability


Select a Size

Change View
10 MG
€358.00

About This Item

Linear Formula:
NH2(CD2)3CH(COOH)(NH2) •HCl
CAS Number:
Molecular Weight:
174.66
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.13

€358.00


Check Cart for Availability

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

powder

mp

245 °C (dec) (lit.)

mass shift

M+6

storage temp.

room temp

SMILES string

Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])[C@H](N)C(O)=O

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1/i1D2,2D2,3D2;

InChI key

GGTYBZJRPHEQDG-IYAGNYDCSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Caitlyn Makins et al.
Biochimica et biophysica acta, 1865(8), 1077-1084 (2017-05-22)
Ornithine 4,5-aminomutase (OAM) from Clostridium sticklandii is an adenosylcobalamin (AdoCbl) and pyridoxal 5'-phosphate (PLP)-dependent enzyme that catalyzes a 1,2-amino shift, interconverting d-ornithine and 2S, 4R-diaminopentanoate. The reaction occurs via a radical-based mechanism whereby a PLP-bound substrate radical undergoes intramolecular isomerization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service