Skip to Content
Merck
All Photos(1)

Key Documents

578886

Sigma-Aldrich

L-Citrulline-4,4,5,5-d4

98 atom % D, 97% (CP)

Synonym(s):

L-Ornithine-4,4,5,5-d4

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€1,070.00

€1,070.00


Check Cart for Availability

Request more information

Select a Size

Change View
10 MG
€1,070.00

About This Item

Empirical Formula (Hill Notation):
C6D4H9N3O3
CAS Number:
Molecular Weight:
179.21
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€1,070.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

97% (CP)

form

solid

mp

216-218 °C (dec.) (lit.)

mass shift

M+4

storage temp.

room temp

SMILES string

[2H]C([2H])(C[C@H](N)C(O)=O)C([2H])([2H])NC(N)=O

InChI

1S/C6H13N3O3/c7-4(5(10)11)2-1-3-9-6(8)12/h4H,1-3,7H2,(H,10,11)(H3,8,9,12)/t4-/m0/s1/i1D2,3D2

InChI key

RHGKLRLOHDJJDR-KKOAYFQZSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter E L Pontoppidan et al.
The Journal of nutrition, 145(7), 1472-1480 (2015-05-29)
Intensive chemotherapy frequently results in gut toxicity, indicated by oral and intestinal mucositis, resulting in poor treatment outcomes and increased mortality. There are no effective preventive strategies against gut toxicity and the role of diet is unknown. We hypothesized that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service