Skip to Content
Merck
All Photos(1)

Key Documents

603511

Sigma-Aldrich

(Trimethylsilyl)acetylene-13C2

99 atom % 13C, 95% (CP)

Synonym(s):

Ethynyl-13C2-trimethylsilane

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€779.00
1 G
€2,100.00

€779.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
€779.00
1 G
€2,100.00

About This Item

Linear Formula:
(CH3)3Si13C≡13CH
CAS Number:
Molecular Weight:
100.20
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

€779.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

95% (CP)

bp

53 °C

mass shift

M+2

storage temp.

2-8°C

SMILES string

C[Si](C)(C)[13C]#[13CH]

InChI

1S/C5H10Si/c1-5-6(2,3)4/h1H,2-4H3/i1+1,5+1

InChI key

CWMFRHBXRUITQE-AEGZSVGQSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-29.2 °F

Flash Point(C)

-34 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service