Skip to Content
Merck
All Photos(1)

Key Documents

723266

Sigma-Aldrich

Dehydroepiandrosterone-2,2,3,4,4,6-d6 sulfate sodium salt

97 atom % D, 98% (CP)

Synonym(s):

Androst-5-en-17-one-2,2,3,4,4,6-d6, 3-(sulfooxy sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€239.00
5 MG
€765.00
10 MG
€1,410.00

€239.00


Estimated to ship on01 June 2025


Request more information

Select a Size

Change View
1 MG
€239.00
5 MG
€765.00
10 MG
€1,410.00

About This Item

Empirical Formula (Hill Notation):
C19D6H21O5NaS
CAS Number:
Molecular Weight:
396.51
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.24

€239.00


Estimated to ship on01 June 2025


Request more information

isotopic purity

97 atom % D

Assay

98% (CP)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@]12C(C([2H])([2H])[C@](OS(=O)(O[Na])=O)([2H])C([2H])([2H])C2)=C([2H])CC3C1CC[C@@]4(C)C3CCC4=O

InChI

1S/C19H28O5S.Na/c1-18-9-7-13(24-25(21,22)23)11-12(18)3-4-14-15-5-6-17(20)19(15,2)10-8-16(14)18;/h3,13-16H,4-11H2,1-2H3,(H,21,22,23);/q;+1/p-1/t13-,14?,15?,16?,18-,19-;/m0./s1/i3D,7D2,11D2,13D;

InChI key

GFJWACFSUSFUOG-WNCZOCEESA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service