Skip to Content
Merck
All Photos(2)

Key Documents

700087P

Avanti

DHEA

Avanti Research - A Croda Brand

Synonym(s):

DHEA; 3β-hydroxy-5-androsten-17-one; 5-androsten-3β-ol-17-one; dehydroisoandrosterone; trans-dehydroandrosterone; Prasterone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€54.50

€54.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€54.50

About This Item

Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€54.50


Check Cart for Availability

Request a Bulk Order

description

Dehydroepiandrosterone

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (700087P-1g)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@](CC3)(C(=O)CC4)C)CC=C2C1)C

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h3,13-16,20H,4-11H2,1-2H3/t13-,14-,15-,16-,18-,19-/m0/s1

InChI key

FMGSKLZLMKYGDP-USOAJAOKSA-N

General description

25-hydroxy-cholesterol is a metabolite of cholesterol. It is produced by macrophages.[1]
DHEA (dehydroepiandrosterone) is an endogenous steroid hormone secreted by the adrenal gland. DHEA serves as precursor to male and female sex hormones (androgens and estrogens).
Dehydroepiandrosterone (DHEA) is a carbon (C)-19 neuroactive steroid. It is synthesized by the adrenal gland in humans and mammals.[2]

Biochem/physiol Actions

25-hydroxy-cholesterol represses the production of IgA by B cells, manages the migration of activated B cells in the germinal follicle. It regulates the differentiation of monocytes into macrophages.[1] 25-hydroxy-cholesterol plays a critical role in lipid biosynthesis and immunity. It inhibits sterol regulatory element-binding protein (SREBP) proteolytic processing and modulates cholesterol metabolism.[3]
Dehydroepiandrosterone (DHEA) regulates neural functions, such as neurogenesis, neuroprotection, neuronal growth and differentiation. It is used to treat multiple sclerosis. DHEA acts as a precursor of sex steroid biosynthesis.[2]

Packaging

20 mL Clear Glass Screw Cap Vial (700087P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

25-Hydroxycholesterol activates the integrated stress response to reprogram transcription and translation in macrophages
Shibata N, et al.
The Journal of biological chemistry, 288(50), 35812-35823 (2013)
25-Hydroxycholesterol: a new life in immunology
McDonald JG and Russell DW
Journal of Leukocyte Biology, 88(6), 1071-1072 (2010)
Joseph S Dillon
Current drug targets. Inflammation and allergy, 4(3), 377-385 (2005-08-17)
Dehydroepiandrosterone (DHEA) and dehydroepiandrosterone sulfate (DHEAS) are metabolic intermediates in the production of potent androgens, estrogens and other less well-characterized steroids. DHEA(S) and closely related steroid hormones have a variety of immunological effects both in vitro and in vivo in
Marco Racchi et al.
CNS drug reviews, 9(1), 21-40 (2003-02-22)
The physiological role of dehydroepiandrosterone (DHEA) and its sulphated ester DHEA(S) has been studied for nearly 2 decades and still eludes final clarification. The major interest in DHEA derives from its unique pattern of activity. Its levels exhibit a dramatic
Sayali Upasham et al.
The Analyst, 145(3), 784-796 (2019-12-24)
Chronobiology is defined as the temporal fluctuations occurring in the human physiology due to the circadian cycle. These fluctuations are good indicators of the functioning of the Hypothalamic-Pituitary-Adrenal axis (HPA axis) and can be tracked by using biomarkers: cortisol and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service