Skip to Content
Merck
All Photos(1)

Key Documents

T-070

Supelco

Testosterone-2,3,4-13C3 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€338.00

€338.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€338.00

About This Item

Empirical Formula (Hill Notation):
13C3C16H28O2
CAS Number:
Molecular Weight:
291.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€338.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=[13CH][13C](=O)[13CH2]C[C@]34C)[C@@H]1CC[C@@H]2O

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1/i7+1,11+1,13+1

InChI key

MUMGGOZAMZWBJJ-KRMLYXHASA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard for monitoring serum testosterone levels in men, women, and children by LC-MS/MS. The Centers for Disease Control (CDC) Hormone Standardization Project cites Testosterone-2,3,4-13C3 as the internal standard of choice for measurement of testosterone at low concentrations in biological fluids by LC-MS/MS. For these analyses, 13C-labeled internal standards can eliminate potential issues of isotopic exchange and minimize separation of later-eluting analytes from a higher substituted deuterium-labeled internal standard. Acetonitrile was chosen as diluent for its ease of handling and miscibility with water and biological specimens.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu Zhang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 965, 183-189 (2014-07-20)
A liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method was developed, using testosterone-d3 as a surrogate analyte, for the simultaneous quantification of goserelin and testosterone in rat plasma. According to this method, the pharmacokinetic and pharmacodynamic data were obtained from

Articles

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service