Skip to Content
Merck
All Photos(1)

Key Documents

710180

Sigma-Aldrich

Di-2-methoxyethyl azodicarboxylate

≥90.0%

Synonym(s):

Azodicarboxylic acid bis(2-methoxyethyl ester), DMEAD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O6
CAS Number:
Molecular Weight:
234.21
Beilstein:
11134057
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90.0%

form

crystals

functional group

azo
ether

storage temp.

2-8°C

SMILES string

COCCOC(=O)\N=N\C(=O)OCCOC

InChI

1S/C8H14N2O6/c1-13-3-5-15-7(11)9-10-8(12)16-6-4-14-2/h3-6H2,1-2H3/b10-9+

InChI key

PGHKJMVOHWKSLJ-MDZDMXLPSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Self-react. D - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Di-2-methoxyethyl azodicarboxylate (DMEAD): an inexpensive and separation-friendly alternative reagent for the Mitsunobu reaction.
Sugimura, T. and Hagiya, K.
Chemistry Letters (Jpn), 36, 566-567 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service