Skip to Content
Merck
All Photos(3)

Key Documents

D3648

Sigma-Aldrich

Diamide

Diamide
1 of 1 reviewers received a sample product or took part in a promotion

Synonym(s):

1,1′-Azobis(N,N-dimethylformamide), N,N,N′,N′-Tetramethylazodicarboxamide, Azodicarboxylic acid bis(dimethylamide), Diazenedicarboxylic acid bis(N,N-dimethylamide), TMAD

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€135.00
5 G
€546.00

€135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€135.00
5 G
€546.00

About This Item

Linear Formula:
(CH3)2NCON=NCON(CH3)2
CAS Number:
Molecular Weight:
172.19
Beilstein:
1910409
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.22

€135.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (TLC)

Quality Level

form

powder

mp

113-115 °C

solubility

water: 19.60-21.00 mg/mL, clear to slightly hazy, orange

storage temp.

−20°C

SMILES string

CN(C)C(=O)\N=N\C(=O)N(C)C

InChI

1S/C6H12N4O2/c1-9(2)5(11)7-8-6(12)10(3)4/h1-4H3/b8-7+

InChI key

VLSDXINSOMDCBK-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reported to be of use as a thiol oxidizing agent. Diamide has been used to titrate protein glutathiolation to discriminate from other oxidative protein modifications. Treatment increased protein glutathiolation in a concentration-dependent manner and had comparably little effect on protein-protein disulfide formation.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bradford G Hill et al.
Biochimica et biophysica acta, 1797(2), 285-295 (2009-11-21)
Protein thiolation by glutathione is a reversible and regulated post-translational modification that is increased in response to oxidants and nitric oxide. Because many mitochondrial enzymes contain critical thiol residues, it has been hypothesized that thiolation reactions regulate cell metabolism and
Carlos A Sorgi et al.
Scientific reports, 7(1), 10981-10981 (2017-09-10)
The differentiation of resident tissue macrophages from embryonic precursors and that of inflammatory macrophages from bone marrow cells leads to macrophage heterogeneity. Further plasticity is displayed through their ability to be polarized as subtypes M1 and M2 in a cell
Dierk-Christoph Pöther et al.
International journal of medical microbiology : IJMM, 303(3), 114-123 (2013-03-23)
Bacillithiol (Cys-GlcN-malate, BSH) serves as a major low molecular weight thiol in low GC Gram-positive bacteria including Bacillus species and a variety of Staphylococcus aureus strains. These bacteria do not produce glutathione (GSH). In this study, HPLC analyses were used
Yong Tao et al.
Insect biochemistry and molecular biology, 43(9), 820-828 (2013-06-29)
Anthranilic diamides, which include the new commercial insecticide, chlorantraniliprole, are an exciting new class of chemistry that target insect ryanodine receptors. These receptors regulate release of stored intracellular calcium and play a critical role in muscle contraction. As with insects
Yumei He et al.
Nucleic acids research, 45(1), 106-114 (2016-09-25)
We describe a Pap1-Oxs1 pathway for diamide-induced disulfide stress in Schizosaccharomyces pombe, where the nucleocytoplasmic HMG protein Oxs1 acts cooperatively with Pap1 to regulate transcription. Oxs1 and Pap1 form a complex when cells are exposed to diamide or Cd that

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service