Skip to Content
Merck
All Photos(2)

Key Documents

694134

Sigma-Aldrich

N-(3-Aminopropyl)piperidine

95%

Synonym(s):

3-Piperidinopropylamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€209.00

€209.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€209.00

About This Item

Empirical Formula (Hill Notation):
C8H18N2
CAS Number:
Molecular Weight:
142.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€209.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.476

density

0.895 g/mL at 25 °C

functional group

amine

SMILES string

NCCCN1CCCCC1

InChI

1S/C8H18N2/c9-5-4-8-10-6-2-1-3-7-10/h1-9H2

InChI key

JMUCXULQKPWSTJ-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • A chemotype that inhibits Botulinum neurotoxin serotype A light chain, P. falciparum Malaria, and Ebola filovirus
  • Membrane targeting antibiotics
  • 5-HT3A receptor antagonists
  • Histamine H3 antagonists
  • Berberine derivatives for stabilization of G-quadruplex DNA and down regulation of oncogene c-myc

Reactant for incorporation of basic side chains into cryptolepine scaffold for antimalarial activity

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

180.0 °F

Flash Point(C)

82.2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary E Gagou et al.
Oncotarget, 5(22), 11381-11398 (2014-11-02)
Unwinding duplex DNA is a critical processing step during replication, repair and transcription. Pif1 are highly conserved non-processive 5'->3' DNA helicases with well-established roles in maintenance of yeast genome stability. However, the function of the sole member of Pif1 family
W Guo et al.
Cell death & disease, 5, e1409-e1409 (2014-09-05)
Btk and Etk/BMX are Tec-family non-receptor tyrosine kinases. Btk has previously been reported to be expressed primarily in B cells and has an important role in immune responses and B-cell malignancies. Etk has been shown previously to provide a strong

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service