Merck
  • Human PIF1 helicase supports DNA replication and cell growth under oncogenic-stress.
Human PIF1 helicase supports DNA replication and cell growth under oncogenic-stress.
Oncotarget (2014-11-02)
Mary E Gagou, Anil Ganesh, Geraldine Phear, Darren Robinson, Eva Petermann, Angela Cox, Mark Meuth
ABSTRACT
Unwinding duplex DNA is a critical processing step during replication, repair and transcription. Pif1 are highly conserved non-processive 5'->3' DNA helicases with well-established roles in maintenance of yeast genome stability. However, the function of the sole member of Pif1 family in humans remains unclear. Human PIF1 is essential for tumour cell viability, particularly during replication stress, but is dispensable in non-cancerous cells and Pif1 deficient mice. Here we report that suppression of PIF1 function slows replication fork rates and increases arrested forks during normal cycling conditions. Importantly, PIF1-dependent replication impediments impair S-phase progression and reduce proliferation rates of RAS oncogene-transformed fibroblasts, where replication fork slowing is exacerbated, but not parental, non-cancerous cells. Disrupted fork movement upon PIF1-depletion does not enhance double-stranded break formation or DNA damage responses but affects resumption of DNA synthesis after prolonged replication inhibitor exposure, accompanied by diminished new origin firing and mainly S-phase entry. Taken together, we characterised a functional role for human PIF1 in DNA replication that becomes important for cell growth under oncogenic stress. Given that oncogenes induce high levels of replication stress during the early stages of tumorigenesis, this function of PIF1 could become critical during cancer development.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pif1
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
SYUIQ-5, ≥98% (HPLC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
N-(3-Aminopropyl)piperidine, 95%
Sigma-Aldrich
Fluoroshield, histology mounting medium
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
DAPI, for nucleic acid staining
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard