Skip to Content
Merck
All Photos(1)

Key Documents

656518

Sigma-Aldrich

2-(Aminomethyl)piperidine

97%

Synonym(s):

(RS)-1-(Piperidin-2-yl)methanamine, 2-Piperidinemethanamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€185.00

€185.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€185.00

About This Item

Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€185.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.4854

density

0.9406 g/mL at 25 °C

functional group

amine

SMILES string

NCC1CCCCN1

InChI

1S/C6H14N2/c7-5-6-3-1-2-4-8-6/h6,8H,1-5,7H2

InChI key

RHPBLLCTOLJFPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for reversible aminal formation

Reactant for synthesis of:
  • Annulated N-heterocyclic carbene ligands
  • M3 Muscarinic receptor antagonists
  • Selective 5-ht5A receptor antagonists
  • Ternary platinum(II) complexes

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

154.9 °F

Flash Point(C)

68.3 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service