Skip to Content
Merck
All Photos(1)

Key Documents

690066

Sigma-Aldrich

6-Bromo-2,2′-bipyridine

97%

Synonym(s):

2-Bromo-6-(2-pyridyl)pyridine, 6-Bromo-2,2′-bipyridyl, 6-Bromo-[2,2′]bipyridinyl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€186.00

€186.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€186.00

About This Item

Empirical Formula (Hill Notation):
C10H7BrN2
CAS Number:
Molecular Weight:
235.08
Beilstein:
130238
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€186.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥96.5% (HPLC)
97%

form

lumps

functional group

bromo

SMILES string

Brc1cccc(n1)-c2ccccn2

InChI

1S/C10H7BrN2/c11-10-6-3-5-9(13-10)8-4-1-2-7-12-8/h1-7H

InChI key

NCRIDSGPLISUEU-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Keller et al.
Dalton transactions (Cambridge, England : 2003), 47(40), 14263-14276 (2018-05-24)
Heteroleptic [Cu(P^P)(N^N)][PF6] complexes, where N^N is a halo-substituted 2,2'-bipyridine (bpy) and P^P is either bis(2-(diphenylphosphino)phenyl)ether (POP) or 4,5-bis(diphenylphosphino)-9,9-dimethylxanthene (xantphos) have been synthesized and investigated. To stabilize the tetrahedral geometry of the copper(i) complexes, the steric demands of the bpy ligands

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service