Skip to Content
Merck
All Photos(1)

Key Documents

513881

Sigma-Aldrich

6,6′-Dibromo-2,2′-dipyridyl

90%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€112.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€112.00

About This Item

Empirical Formula (Hill Notation):
C10H6Br2N2
CAS Number:
Molecular Weight:
313.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€112.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

90%

mp

220-223 °C (lit.)

functional group

bromo

SMILES string

Brc1cccc(n1)-c2cccc(Br)n2

InChI

1S/C10H6Br2N2/c11-9-5-1-3-7(13-9)8-4-2-6-10(12)14-8/h1-6H

InChI key

WZVWSOXTTOJQQQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and temperature-dependent conformational preferences of macrocycles containing the 2, 2'-bipyridyl moiety.
Newkome GR, et al.
Journal of the American Chemical Society, 101(16), 4472-4477 (1979)
Binitendra Naath Mongal et al.
Dalton transactions (Cambridge, England : 2003), 48(27), 10070-10077 (2019-06-11)
In this work, two new bis-(hydroxyphenyl)bipyridine based ruthenium complexes with 4-picoline (coded as MCS-B4M) and isonicotinic acid (coded as MCS-B5M), which act as ancillary ligands, have been synthesized and employed for the first time as photosensitizers in photocatalytic hydrogen evolution
Sarah Keller et al.
Dalton transactions (Cambridge, England : 2003), 47(40), 14263-14276 (2018-05-24)
Heteroleptic [Cu(P^P)(N^N)][PF6] complexes, where N^N is a halo-substituted 2,2'-bipyridine (bpy) and P^P is either bis(2-(diphenylphosphino)phenyl)ether (POP) or 4,5-bis(diphenylphosphino)-9,9-dimethylxanthene (xantphos) have been synthesized and investigated. To stabilize the tetrahedral geometry of the copper(i) complexes, the steric demands of the bpy ligands
Ken T Ngo et al.
Journal of the American Chemical Society, 139(7), 2604-2618 (2017-01-25)
Electrocatalytic reduction of CO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service