Skip to Content
Merck
All Photos(2)

Key Documents

802425

Sigma-Aldrich

2-Bromo-1,10-phenanthroline

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€136.00

€136.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€136.00

About This Item

Empirical Formula (Hill Notation):
C12H7BrN2
CAS Number:
Molecular Weight:
259.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€136.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mp

160-165 °C

functional group

bromo

SMILES string

BrC1=NC2=C(N=CC=C3)C3=CC=C2C=C1

InChI

1S/C12H7BrN2/c13-10-6-5-9-4-3-8-2-1-7-14-11(8)12(9)15-10/h1-7H

InChI key

ZRJUDAZGVGIDLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Brominated phenanthroline can be synthesized via a method reported by Baran and coworkers using a two-step protocol through N-Oxide formation using m-CBPA (Aldrich 273031) and bromination using TBAB (Sigma-Aldrich 426288) and p-Toluenesulfonic anhydride (Aldrich 259764).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service