Skip to Content
Merck
All Photos(2)

Documents

681113

Sigma-Aldrich

2-Bromo-4-chlorobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(Br)CO2H
CAS Number:
Molecular Weight:
235.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

157-161 °C

SMILES string

OC(=O)c1ccc(Cl)cc1Br

InChI

1S/C7H4BrClO2/c8-6-3-4(9)1-2-5(6)7(10)11/h1-3H,(H,10,11)

InChI key

USMQLFCVCDEXAK-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service