Skip to Content
Merck
All Photos(1)

Key Documents

666181

Sigma-Aldrich

1,3-Dicyclohexylimidazolium tetrafluoroborate salt

97%

Synonym(s):

1,3-Bis(cyclohexyl)imidazolium tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€453.00

€453.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€453.00

About This Item

Empirical Formula (Hill Notation):
C15H25BF4N2
CAS Number:
Molecular Weight:
320.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€453.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: catalyst

mp

171-175 °C

SMILES string

F[B-](F)(F)F.C1CCC(CC1)n2cc[n+](c2)C3CCCCC3

InChI

1S/C15H25N2.BF4/c1-3-7-14(8-4-1)16-11-12-17(13-16)15-9-5-2-6-10-15;2-1(3,4)5/h11-15H,1-10H2;/q+1;-1

InChI key

CQHXJIHJFMBBQA-UHFFFAOYSA-N

Application

Used as organocatalyst in the catalytic boron conjugate additions to cyclic and acyclic unsaturated carbonyls[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kang-sang Lee et al.
Journal of the American Chemical Society, 131(21), 7253-7255 (2009-05-13)
Metal-free nucleophilic activation of a B-B bond has been exploited in the development of a highly efficient method for conjugate additions of commercially available bis(pinacolato)diboron to cyclic or acyclic alpha,beta-unsaturated carbonyls. The reactions are readily catalyzed by a simple N-heterocyclic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service