Skip to Content
Merck
All Photos(3)

Key Documents

748382

Sigma-Aldrich

Dimethyltin oxide

Synonym(s):

Dimethyloxostannane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€245.00

€245.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€245.00

About This Item

Empirical Formula (Hill Notation):
C2H6OSn
CAS Number:
Molecular Weight:
164.78
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€245.00


Check Cart for Availability

Request a Bulk Order

form

solid

mp

380-390 °C

storage temp.

2-8°C

SMILES string

C[Sn](C)=O

InChI

1S/2CH3.O.Sn/h2*1H3;;

InChI key

WNVQCJNZEDLILP-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service