Skip to Content
Merck
All Photos(1)

Key Documents

538019

Sigma-Aldrich

2-Diphenylphosphinoethyl-functionalized silica gel

200-400 mesh, extent of labeling: 0.7 mmol/g loading

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€408.00

€408.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€408.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€408.00


Check Cart for Availability

Request a Bulk Order

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reaction type: solution phase peptide synthesis
reagent type: ligand

Quality Level

extent of labeling

0.7 mmol/g loading

surface area

500 m2/g

particle size

200-400 mesh

pore size

60 Å pore size

functional group

phosphine

storage temp.

2-8°C

SMILES string

O=[SiH2].CCP(c1ccccc1)c2ccccc2

InChI

1S/C14H15P.H2OSi/c1-2-15(13-9-5-3-6-10-13)14-11-7-4-8-12-14;1-2/h3-12H,2H2,1H3;2H2

InChI key

PWUAVLQPJXAPLL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Functionalized silica gel is used in size-exclusion chromatography of cationic polymers and in the separation of sugars, steroids, cholesterol, and triglycerides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service