Skip to Content
Merck
All Photos(1)

Key Documents

552658

Sigma-Aldrich

3-Bromopropyl-functionalized silica gel

200-400 mesh, extent of labeling: 1.5 mmol/g loading

Synonym(s):

Bromide-3 functionalized silica gel

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€183.00

€183.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€183.00

About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

€183.00


Check Cart for Availability

Request a Bulk Order

extent of labeling

1.5 mmol/g loading

surface area

500 m2/g

particle size

200-400 mesh

pore size

60 Å pore size

SMILES string

O=[SiH2].CCCBr

InChI

1S/C3H7Br.H2OSi/c1-2-3-4;1-2/h2-3H2,1H3;2H2

InChI key

MHCVEVAMXYXCCC-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service