Skip to Content
Merck
All Photos(1)

Key Documents

522317

Sigma-Aldrich

2-Mercapto-4-methyl-5-thiazoleacetic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€135.00

€135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€135.00

About This Item

Empirical Formula (Hill Notation):
C6H7NO2S2
CAS Number:
Molecular Weight:
189.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€135.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

207 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

Cc1nc(S)sc1CC(O)=O

InChI

1S/C6H7NO2S2/c1-3-4(2-5(8)9)11-6(10)7-3/h2H2,1H3,(H,7,10)(H,8,9)

InChI key

KYBOCQHDFLVQIB-UHFFFAOYSA-N

General description

2-Mercapto-4-methyl-5-thiazoleacetic acid is a thiazole derivative. It participates in the preparation of dinuclear [Au-(O4NCS2)]2 and tetranuclear [Au(SSCOOH)]4 complexes.[1]

Application

2-Mercapto-4-methyl-5-thiazoleacetic acid may be used as a capping agent in the preparation of fused spherical gold nanoparticles.[2] It may also be used in the synthesis of eight-tin macrocyclic complex (a 40-membered macrocycle containing two distannoxane ladders).[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chunlin Ma et al.
Dalton transactions (Cambridge, England : 2003), (12)(12), 1785-1786 (2004-09-24)
The self-assembly reaction of 2-mercapto-4-methyl-5-thiazoleacetic acid, sodium methoxide and dimethyltin dichloride in moist dichloromethane affords a novel eight-tin macrocyclic complex, which is a 40-membered macrocycle containing two distannoxane ladders.
Chunlin Ma et al.
Inorganic chemistry, 47(16), 7060-7061 (2008-07-16)
The reaction of 1-(4-carboxyphenyl)-5-mercapto-1 H-tetrazole with trimethyltin chloride in the presence of sodium ethoxide in benzene affords a novel 2D organotin network structure complex 1, which is an unusual organotin network containing a hexanuclear 60-membered organotin macrocycle. The complex has
P Kannan et al.
Analytica chimica acta, 663(2), 158-164 (2010-03-09)
We are reporting the highly sensitive determination of hydroxylamine (HA) using 2-mercapto-4-methyl-5-thiazoleacetic acid (TAA) capped fused spherical gold nanoparticles (AuNPs) modified Au electrode. The fused TAA-AuNPs were immobilized on (3-mercaptopropyl)-trimethoxysilane (MPTS) sol-gel film, which was pre-assembled on Au electrode. The
Dan Zhu et al.
Theranostics, 8(4), 941-954 (2018-02-22)
The recent proposition to combine liposomes with nanoparticles presents great opportunities to develop multifunctional drug delivery platforms. Although impressive progress has been made, attempts to elucidate the role nanoparticles play in the integral nanohybrids are still rather limited. Here, using
Isabel Freitag et al.
Journal of biomedical optics, 20(5), 55002-55002 (2015-05-06)
Identification of tumor and normal cells is a promising application of Raman spectroscopy. The throughput of Raman-assisted cell sorting is limited by low sensitivity. Surface-enhanced Raman spectroscopy (SERS) is a well-recognized candidate to increase the intensity of Raman signals of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service