Skip to Content
Merck
All Photos(1)

Key Documents

407003

Sigma-Aldrich

2-[2-(2-Methoxyethoxy)ethoxy]acetic acid

technical grade

Synonym(s):

O-[2-(2-Methoxyethoxy)ethyl]glycolic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€156.00

€156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€156.00

About This Item

Linear Formula:
CH3(OCH2CH2)2OCH2CO2H
CAS Number:
Molecular Weight:
178.18
Beilstein:
1769137
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€156.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

140 °C/2 mmHg (lit.)

density

1.161 g/mL at 25 °C (lit.)

SMILES string

COCCOCCOCC(O)=O

InChI

1S/C7H14O5/c1-10-2-3-11-4-5-12-6-7(8)9/h2-6H2,1H3,(H,8,9)

InChI key

YHBWXWLDOKIVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Matched Ligands for Small, Stable Colloidal Nanoparticles of Copper, Cuprous Oxide, and Cuprous Sulfide.: This study explores the synthesis and stabilization of colloidal nanoparticles using various ligands, highlighting the potential of 2-[2-(2-Methoxyethoxy)ethoxy]acetic acid in nanoparticle stabilization and applications in biocompatible systems (Cowie BE et al., 2023).

  • Interaction of Surface-Modified Alumina Nanoparticles and Surfactants at an Oil/Water Interface: A Neutron Reflectometry, Scattering, and Enhanced Oil Recovery Study.: This research delves into the surface modifications of nanoparticles to enhance their performance in oil recovery processes, with implications for the use of 2-[2-(2-Methoxyethoxy)ethoxy]acetic acid as a surfactant modifier (Al-Shatty W et al., 2022).

  • Investigations on the Processing of Ceramic Filled Inks for 3D InkJet Printing.: This paper discusses the formulation of ceramic-filled inks for 3D printing, emphasizing the role of 2-[2-(2-Methoxyethoxy)ethoxy]acetic acid in improving the printability and stability of the inks (Graf D et al., 2020).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

351.5 °F - closed cup

Flash Point(C)

177.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haoyu Liu et al.
Carbohydrate polymers, 113, 480-489 (2014-09-27)
Polymer design and selection are crucial to the development of amorphous solid dispersions (ASD) for solubilization of otherwise poorly water-soluble drugs. The matrix polymer is required to interact strongly at the molecular level with the drug to prevent recrystallization, but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service